N-(3-Formyl-5-methylpyridin-2-yl)pivalamide ≥97%

Supplier: AOBChem USA

Warning

52438-1G 52438-500MG 52438-5G
76943-044EA 593.05 USD
76943-044 76943-046 76943-048
N-(3-Formyl-5-methylpyridin-2-yl)pivalamide ≥97%
N-(3-Formyl-5-methylpyridin-2-yl)pivalamide
Formula: C₁₂H₁₆N₂O₂
MW: 220.27 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13176641
CAS Number: 127446-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR