Methyl benzimidate ≥97%

Supplier: AOBChem USA

Warning

52136-1G 52136-25G 52136-500MG 52136-5G
76943-030EA 379.81 USD
76943-030 76943-032 76943-034 76943-036
Methyl benzimidate ≥97%
Methyl benzimidate
Formula: C₈H₉NO
MW: 135.163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02728616
CAS Number: 7471-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR