Ethyl 2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate ≥97%

Supplier: AOBChem USA

Warning

52070-1G 52070-25G 52070-500MG 52070-5G
76943-014EA 334.19 USD
76943-014 76943-016 76943-018 76943-020
Ethyl 2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate ≥97%
Ethyl 2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Formula: C₁₅H₂₀BFO₄
Storage Temperature: Ambient
MDL Number: MFCD18383987
CAS Number: 1198615-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR