Methyl 3-bromo-5-chloroisonicotinate ≥97%

Supplier: AOBChem USA

Warning

51798-1G 51798-25G 51798-500MG 51798-5G
76942-998EA 227.04 USD
76942-998 76943-000 76943-002 76943-004
Methyl 3-bromo-5-chloroisonicotinate ≥97%
Methyl 3-bromo-5-chloroisonicotinate
Formula: C₇H₅BrClNO₂
Storage Temperature: Ambient
MDL Number: MFCD14698106
CAS Number: 1214361-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR