6-Iodo-1,4-benzodioxane ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 6-Iodo-2,3-dihydro-1,4-benzodioxine, 1,2-Ethylenedioxy-4-iodobenzene, 6-Iodo-2,3-dihydrobenzo[b][1,4]dioxine

51698-25G 51698-5G
76942-986EA 254.62 USD
76942-986 76942-988
6-Iodo-1,4-benzodioxane ≥97% research grade
6-Iodo-1,4-benzodioxane
Formula: C₈H₇IO₂
MW: 262.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00221460
CAS Number: 57744-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR