Methyl dibenzo[b,d]thiophene-4-carboxylate ≥97%

Supplier: AOBChem USA

Warning

49680-25G 49680-5G
76942-900EA 453.01 USD
76942-900 76942-902
Methyl dibenzo[b,d]thiophene-4-carboxylate ≥97%
Methyl dibenzo[b,d]thiophene-4-carboxylate
Formula: C₁₄H₁₀O₂S
MW: 242.293 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00092758
CAS Number: 60718-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR