Ethyl 4-methoxy-2,3-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

49419-1G 49419-25G 49419-500MG 49419-5G
76942-884EA 334.19 USD
76942-884 76942-886 76942-888 76942-890
Ethyl 4-methoxy-2,3-dimethylbenzoate ≥97%
Ethyl 4-methoxy-2,3-dimethylbenzoate
Formula: C₁₂H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD6204629
CAS Number: 128209-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR