Methyl 5-bromo-2-iodo-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

49167-1G 49167-25G 49167-500MG 49167-5G
76942-860EA 292.81 USD
76942-860 76942-862 76942-864 76942-866
Methyl 5-bromo-2-iodo-3-methylbenzoate ≥97%
Methyl 5-bromo-2-iodo-3-methylbenzoate
Formula: C₉H₈BrIO₂
MW: 354.967 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18379816
CAS Number: 1263376-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR