Ethyl-2,4-dichlorobenzoate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2,4-dichlorobenzoate

49116-25G 49116-5G
76942-850EA 233.4 USD
76942-850 76942-852
Ethyl-2,4-dichlorobenzoate ≥97%
Ethyl-2,4-dichlorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈Cl₂O₂
MW: 219.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00277702
CAS Number: 56882-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR