Ethyl 4-isopropoxy-3-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

48620-1G 48620-25G 48620-500MG 48620-5G
76942-780EA 213.25 USD
76942-780 76942-782 76942-784 76942-786
Ethyl 4-isopropoxy-3-methoxybenzoate ≥97%
Ethyl 4-isopropoxy-3-methoxybenzoate
Formula: C₁₃H₁₈O₄
Storage Temperature: Refrigerator
MDL Number: MFCD27964757
CAS Number: 199102-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR