Ethyl 5-bromo-2-chloro-4-fluorobenzoate ≥97% research grade

Supplier: AOBChem USA

Warning

48502-1G 48502-25G 48502-500MG 48502-5G
76942-764EA 409.51 USD
76942-764 76942-766 76942-768 76942-770
Ethyl 5-bromo-2-chloro-4-fluorobenzoate ≥97% research grade
Ethyl 5-bromo-2-chloro-4-fluorobenzoate
Formula: C₉H₇BrClFO₂
MW: 281.506 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24107048
CAS Number: 351325-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR