Methyl 2-fluoro-5-methylisonicotinate 97 Research

Supplier: AOBChem USA

Warning

48409-1G 48409-500MG 48409-5G
76942-754EA 694.89 USD
76942-754 76942-756 76942-758
Methyl 2-fluoro-5-methylisonicotinate 97 Research
Methyl 2-fluoro-5-methylisonicotinate
Formula: C₈H₈FNO₂
MW: 169.153 g/mol
Storage Temperature: Ambient
CAS Number: 1227516-39-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR