Methyl 4-bromo-2-methylsulfanylbenzoate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 4-bromo-2-(methylthio)benzoate

48115-1G 48115-500MG 48115-5G
76942-734EA 720.36 USD
76942-734 76942-736 76942-738
Methyl 4-bromo-2-methylsulfanylbenzoate ≥97%
Methyl 4-bromo-2-methylsulfanylbenzoate

Precaución: for Research Use Only

Formula: C₉H₉BrO₂S
MW: 261.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22488927
CAS Number: 1824127-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR