Ethyl 2,6-difluoro-3-iodobenzoate ≥95%

Supplier: AOBChem USA

Warning

47961-1G 47961-25G 47961-500MG 47961-5G
76942-720EA 155.96 USD
76942-720 76942-722 76942-724 76942-726
Ethyl 2,6-difluoro-3-iodobenzoate ≥95%
Ethyl 2,6-difluoro-3-iodobenzoate
Formula: C₉H₇F₂IO₂
MW: 312.052 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24107057
CAS Number: 1520299-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR