Benzofuran-2-carboxaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

47835-25G 47835-5G
76942-708EA 215.37 USD
76942-708 76942-710
Benzofuran-2-carboxaldehyde ≥97% research grade
Benzofuran-2-carboxaldehyde
Formula: C₉H₆O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00015463
CAS Number: 4265-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR