Methyl 3-chloro-2-fluoro-5-iodobenzoate ≥97%

Supplier: AOBChem USA

Warning

47269-1G 47269-25G 47269-500MG 47269-5G
76942-684EA 352.22 USD
76942-684 76942-686 76942-688 76942-690
Methyl 3-chloro-2-fluoro-5-iodobenzoate ≥97%
Methyl 3-chloro-2-fluoro-5-iodobenzoate
Formula: C₈H₅ClFIO₂
Storage Temperature: Refrigerator
MDL Number: MFCD24107277
CAS Number: 1537505-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR