Ethyl 5-bromo-3-chloro-2-fluorobenzoate ≥95% research grade

Supplier: AOBChem USA

Warning

47259-1G 47259-25G 47259-500MG 47259-5G
76942-668EA 379.81 USD
76942-668 76942-670 76942-672 76942-674
Ethyl 5-bromo-3-chloro-2-fluorobenzoate ≥95% research grade
Ethyl 5-bromo-3-chloro-2-fluorobenzoate
Formula: C₉H₇BrClFO₂
Storage Temperature: Ambient
MDL Number: MFCD24107278
CAS Number: 1520957-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR