Methyl 5-bromo-4-fluoro-2-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

46876-1G 46876-25G 46876-500MG 46876-5G
76942-654EA 138.98 USD
76942-654 76942-656 76942-658 76942-660
Methyl 5-bromo-4-fluoro-2-methoxybenzoate ≥97%
Methyl 5-bromo-4-fluoro-2-methoxybenzoate
Formula: C₉H₈BrFO₃
MW: 263.604 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16036943
CAS Number: 314298-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR