Benzyl 5-bromopyridine-2-carboxylate ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: Benzyl 5-bromopicolinate

46651-1G 46651-25G 46651-500MG 46651-5G
76942-638EA 124.13 USD
76942-638 76942-640 76942-642 76942-644
Benzyl 5-bromopyridine-2-carboxylate ≥97% research grade
Benzyl 5-bromopyridine-2-carboxylate
Formula: C₁₃H₁₀BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD11110600
CAS Number: 188052-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR