Methyl 2-bromo-3-fluoroisonicotinate ≥97%

Supplier: AOBChem USA

Warning

46231-1G 46231-25G 46231-500MG 46231-5G
76942-598EA 254.62 USD
76942-598 76942-600 76942-602 76942-604
Methyl 2-bromo-3-fluoroisonicotinate ≥97%
Methyl 2-bromo-3-fluoroisonicotinate
Formula: C₇H₅BrFNO₂
MW: 234.023 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698109
CAS Number: 1214385-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR