Methyl 2-chloro-4-fluoro-3-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

46083-1G 46083-25G 46083-500MG 46083-5G
76942-560EA 260.99 USD
76942-560 76942-562 76942-564 76942-566
Methyl 2-chloro-4-fluoro-3-methoxybenzoate ≥97%
Methyl 2-chloro-4-fluoro-3-methoxybenzoate
Formula: C₉H₈ClFO₃
MW: 218.609 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707153
CAS Number: 2386559-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR