Methyl 4-methylthiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(Methoxycarbonyl)-4-methylthiophene, Methyl 4-methyl-2-thenoate

45949-25G 45949-5G
76942-520EA 268.41 USD
76942-520 76942-522
Methyl 4-methylthiophene-2-carboxylate ≥97%
Methyl 4-methylthiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈O₂S
MW: 156.202 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01763641
CAS Number: 28686-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR