Methyl 3-hydroxy-5-methoxybenzoate ≥97% research grade

Supplier: AOBChem USA

Warning

45661-1G 45661-25G 45661-500MG 45661-5G
76942-448EA 206.88 USD
76942-448 76942-450 76942-452 76942-454
Methyl 3-hydroxy-5-methoxybenzoate ≥97% research grade
Methyl 3-hydroxy-5-methoxybenzoate
Formula: C₉H₁₀O₄
MW: 182.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00601487
CAS Number: 19520-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR