7-Bromo-3-iodo-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

45562-1G 45562-25G 45562-5G
76942-428EA 99.73 USD
76942-428 76942-430 76942-432
7-Bromo-3-iodo-1H-indazole ≥97%
7-Bromo-3-iodo-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrIN₂
MW: 322.9285 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08689979
CAS Number: 944904-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR