6-Propoxy-2-naphthoic acid ≥97%

Supplier: AOBChem USA

Warning

45541-1G 45541-25G 45541-500MG 45541-5G
76942-412EA 126.25 USD
76942-412 76942-414 76942-416 76942-418
6-Propoxy-2-naphthoic acid ≥97%
6-Propoxy-2-naphthoic acid
Formula: C₁₄H₁₄O₃
MW: 230.259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707123
CAS Number: 561013-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR