Ethyl 4-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate ≥97%

Supplier: AOBChem USA

Warning

45493-1G 45493-25G 45493-5G
76942-398EA 101.85 USD
76942-398 76942-400 76942-402
Ethyl 4-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate ≥97%
Ethyl 4-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Formula: C₁₅H₂₀BClO₄
Storage Temperature: Refrigerator
MDL Number: MFCD28148050
CAS Number: 1523239-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR