Methyl 4-chloro-3-fluoro-2-methoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

38779-1G 38779-500MG 38779-5G
76942-372EA 676.86 USD
76942-372 76942-374 76942-376
Methyl 4-chloro-3-fluoro-2-methoxybenzoate ≥95%
Methyl 4-chloro-3-fluoro-2-methoxybenzoate
Formula: C₉H₈ClFO₃
Storage Temperature: Ambient
CAS Number: 2114129-19-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR