Methyl 3,6-difluoro-2-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

38715-1G 38715-25G 38715-500MG 38715-5G
76942-346EA 266.29 USD
76942-346 76942-348 76942-350 76942-352
Methyl 3,6-difluoro-2-methoxybenzoate ≥97%
Methyl 3,6-difluoro-2-methoxybenzoate
Formula: C₉H₈F₂O₃
MW: 202.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18399130
CAS Number: 1261830-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR