6-Ethoxy-2-fluoro-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

38643-1G 38643-25G 38643-500MG 38643-5G
76942-298EA 346.92 USD
76942-298 76942-300 76942-302 76942-304
6-Ethoxy-2-fluoro-3-methylbenzaldehyde ≥97%
6-Ethoxy-2-fluoro-3-methylbenzaldehyde
Formula: C₁₀H₁₁FO₂
MW: 182.192 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD33403788
CAS Number: 2385877-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR