Methyl 3-fluoro-4-isopropoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

38620-1G 38620-25G 38620-5G
76942-284EA 88.06 USD
76942-284 76942-286 76942-288
Methyl 3-fluoro-4-isopropoxybenzoate ≥97%
Methyl 3-fluoro-4-isopropoxybenzoate
Formula: C₁₁H₁₃FO₃
MW: 212.218 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23115009
CAS Number: 1346498-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR