Methyl 6-chloro-4-iodonicotinate ≥97%

Supplier: AOBChem USA

Warning

38550-1G 38550-500MG 38550-5G
76942-256EA 973.91 USD
76942-256 76942-258 76942-260
Methyl 6-chloro-4-iodonicotinate ≥97%
Methyl 6-chloro-4-iodonicotinate
Formula: C₇H₅ClINO₂
Storage Temperature: Refrigerator
MDL Number: MFCD22570686
CAS Number: 1824572-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR