Methyl 2-(benzyloxy)-4,5-dimethylbenzoate ≥95%

Supplier: AOBChem USA

Warning

38509-1G 38509-500MG 38509-5G
76942-246EA 498.63 USD
76942-246 76942-248 76942-250
Methyl 2-(benzyloxy)-4,5-dimethylbenzoate ≥95%
Methyl 2-(benzyloxy)-4,5-dimethylbenzoate
Formula: C₁₇H₁₈O₃
MW: 270.323 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707095
CAS Number: 2432848-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR