Ethyl 4-chloro-3-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

38373-1G 38373-25G 38373-500MG 38373-5G
76942-208EA 138.98 USD
76942-208 76942-210 76942-212 76942-214
Ethyl 4-chloro-3-(trifluoromethyl)benzoate ≥97%
Ethyl 4-chloro-3-(trifluoromethyl)benzoate
Formula: C₁₀H₈ClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD06204386
CAS Number: 773134-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR