Ethyl 3-chloro-2-methoxyisonicotinate ≥97%

Supplier: AOBChem USA

Warning

38329-1G 38329-25G 38329-500MG 38329-5G
76942-180EA 274.78 USD
76942-180 76942-182 76942-184 76942-186
Ethyl 3-chloro-2-methoxyisonicotinate ≥97%
Ethyl 3-chloro-2-methoxyisonicotinate
Formula: C₉H₁₀ClNO₃
MW: 215.634 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14698368
CAS Number: 1214347-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR