Methyl-4-methoxy-2-methylbenzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
38311-25G
38311-5G
76942-170EA
283.27
USD
76942-170
76942-172
Methyl-4-methoxy-2-methylbenzoate ≥97%
Methyl-4-methoxy-2-methylbenzoate
Formula:
C₁₀H₁₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD06203918 CAS Number: 35598-05-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...