Ethyl 4-(benzyloxy)-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

38256-1G 38256-25G 38256-500MG 38256-5G
76942-154EA 296 USD
76942-154 76942-156 76942-158 76942-160
Ethyl 4-(benzyloxy)-2-methylbenzoate ≥97%
Ethyl 4-(benzyloxy)-2-methylbenzoate
Formula: C₁₇H₁₈O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00598356
CAS Number: 1256482-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR