N-Cyclopropyl-3-fluoro-2-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

38238-1G 38238-25G 38238-5G
76942-138EA 76.39 USD
76942-138 76942-140 76942-142
N-Cyclopropyl-3-fluoro-2-methylbenzamide ≥97%
N-Cyclopropyl-3-fluoro-2-methylbenzamide
Formula: C₁₁H₁₂FNO
MW: 193.217 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32632321
CAS Number: 1934600-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR