N-BOC-4-bromo-3-methylaniline ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: tert-Butyl (4-bromo-3-methylphenyl)carbamate

38183-25G 38183-5G
76942-128EA 595.17 USD
76942-128 76942-130
N-BOC-4-bromo-3-methylaniline ≥97% research grade
N-BOC-4-bromo-3-methylaniline
Formula: C₁₂H₁₆BrNO₂
MW: 286.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21170329
CAS Number: 654056-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR