Methyl 4,6-dibromo-2-fluoro-3-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

38159-1G 38159-25G 38159-500MG 38159-5G
76942-098EA 201.58 USD
76942-098 76942-100 76942-102 76942-104
Methyl 4,6-dibromo-2-fluoro-3-methoxybenzoate ≥97%
Methyl 4,6-dibromo-2-fluoro-3-methoxybenzoate
Formula: C₉H₇Br₂FO₃
MW: 341.956 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32707069
CAS Number: 2239325-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR