Methyl 4-bromo-2-fluoro-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

38101-1G 38101-25G 38101-500MG 38101-5G
76942-084EA 227.04 USD
76942-084 76942-086 76942-088 76942-090
Methyl 4-bromo-2-fluoro-3-methylbenzoate ≥97%
Methyl 4-bromo-2-fluoro-3-methylbenzoate
Formula: C₉H₈BrFO₂
MW: 247.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13184289
CAS Number: 1206680-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR