Ethyl 6-bromo-2-chloro-3-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

38093-1G 38093-25G 38093-500MG 38093-5G
76942-068EA 213.25 USD
76942-068 76942-070 76942-072 76942-074
Ethyl 6-bromo-2-chloro-3-methoxybenzoate ≥97%
Ethyl 6-bromo-2-chloro-3-methoxybenzoate
Formula: C₁₀H₁₀BrClO₃
Storage Temperature: Refrigerator
MDL Number: MFCD27920753
CAS Number: 1478503-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR