Ethyl 3-bromo-5-chlorobenzoate ≥95%

Supplier: AOBChem USA

Warning

37949-1G 37949-25G 37949-5G
76942-022EA 102.91 USD
76942-022 76942-024 76942-026
Ethyl 3-bromo-5-chlorobenzoate ≥95%
Ethyl 3-bromo-5-chlorobenzoate
Formula: C₉H₈BrClO₂
Storage Temperature: Ambient
MDL Number: MFCD10566220
CAS Number: 1095274-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR