Methyl 4-(benzyloxy)-5-bromo-2,3-difluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

37887-1G 37887-500MG 37887-5G
76942-010EA 653.52 USD
76942-010 76942-012 76942-014
Methyl 4-(benzyloxy)-5-bromo-2,3-difluorobenzoate ≥97%
Methyl 4-(benzyloxy)-5-bromo-2,3-difluorobenzoate
Formula: C₁₅H₁₁BrF₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD32632271
CAS Number: 2404733-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR