Ethyl 3-methoxy-4-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

37765-25G 37765-5G
76942-000EA 346.92 USD
76942-000 76942-002
Ethyl 3-methoxy-4-methylbenzoate ≥97%
Ethyl 3-methoxy-4-methylbenzoate
Formula: C₁₁H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD12025371
CAS Number: 86239-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR