Methyl 5-hexylthiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

37662-1G 37662-25G 37662-5G
76941-986EA 79.57 USD
76941-986 76941-988 76941-990
Methyl 5-hexylthiophene-2-carboxylate ≥97%
Methyl 5-hexylthiophene-2-carboxylate
Formula: C₁₂H₁₈O₂S
Storage Temperature: Refrigerator
MDL Number: MFCD18415011
CAS Number: 74965-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR