Methyl 5-butylthiophene-2-carboxylate ≥97%
Supplier: AOBChem USA
|
Warning
|
37660-1G
37660-25G
37660-5G
76941-972EA
79.57
USD
76941-972
76941-974
76941-976
Methyl 5-butylthiophene-2-carboxylate ≥97%
Methyl 5-butylthiophene-2-carboxylate
Formula:
C₁₀H₁₄O₂S Storage Temperature: Refrigerator |
MDL Number:
MFCD32632253 CAS Number: 161618-23-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...