Methyl 2-fluoro-3-hydroxybenzoate ≥97% research grade

Supplier: AOBChem USA

Warning

37608-1G 37608-25G 37608-500MG 37608-5G
76941-928EA 135.8 USD
76941-928 76941-930 76941-932 76941-934
Methyl 2-fluoro-3-hydroxybenzoate ≥97% research grade
Methyl 2-fluoro-3-hydroxybenzoate
Formula: C₈H₇FO₃
Storage Temperature: Refrigerator
MDL Number: MFCD14698042
CAS Number: 1214324-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR