Ethyl 3,4-dichloro-2-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

37605-1G 37605-500MG 37605-5G
76941-914EA 589.87 USD
76941-914 76941-916 76941-918
Ethyl 3,4-dichloro-2-fluorobenzoate ≥97%
Ethyl 3,4-dichloro-2-fluorobenzoate
Formula: C₉H₇Cl₂FO₂
MW: 237.055 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28786002
CAS Number: 1806298-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR