tert-Butyl 2-bromo-5-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

37586-25G 37586-5G
76941-906EA 466.8 USD
76941-906 76941-908
tert-Butyl 2-bromo-5-(trifluoromethyl)benzoate ≥97%
tert-Butyl 2-bromo-5-(trifluoromethyl)benzoate
Formula: C₁₂H₁₂BrF₃O₂
MW: 325.122 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23598300
CAS Number: 1515745-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR