Methyl 2,3-dichloro-6-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

37575-1G 37575-25G 37575-5G
76941-886EA 84.88 USD
76941-886 76941-888 76941-890
Methyl 2,3-dichloro-6-fluorobenzoate ≥97%
Methyl 2,3-dichloro-6-fluorobenzoate
Formula: C₈H₅Cl₂FO₂
MW: 223.029 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14698029
CAS Number: 1214386-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR